Skip to Content
MilliporeSigma
Brilliant blue FCF certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Brilliant Blue FCF

Brilliant blue FCF certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Erioglaucine disodium salt, Acid Blue 9, Alphazurine FG, FD&C;BLUE No. 1
Empirical Formula (Hill Notation):
C37H34Na2N2O9S3
CAS Number:
Molecular Weight:
792.85

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique