![Benzoic acid European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/358/205/9c533261-2b7a-424b-81ae-206f0849e268/640/9c533261-2b7a-424b-81ae-206f0849e268.png)
Benzoic acid
![Benzoic acid European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/358/205/9c533261-2b7a-424b-81ae-206f0849e268/640/9c533261-2b7a-424b-81ae-206f0849e268.png)
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2