Skip to Content
MilliporeSigma
Ammonium phosphate dibasic SAJ first grade, 98.5-101.0%

Ammonium phosphate dibasic

Ammonium phosphate dibasic SAJ first grade, 98.5-101.0%
Synonym(s):
Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)
Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Application
Special Grade
Technique