<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Ammonium oxalate monohydrate
Synonym(s):
Oxalic acid diammonium salt
Linear Formula:
(NH
4
)
2
C
2
O
4
· H
2
O
CAS Number:
6009-70-7
Molecular Weight:
142.11
EC Number:
238-135-4
Products
Shipping
Ships Today (2)
Product Category
building blocks (2)
salts (2)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
221716
Ammonium oxalate monohydrate,
ACS reagent, ≥99%
Expand
View Pricing
32304
Ammonium oxalate monohydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-101.0%
Expand
View Pricing
379743
Ammonium oxalate monohydrate,
≥99.99% trace metals basis
Expand
View Pricing
09898
Ammonium oxalate monohydrate,
BioUltra, ≥99.5% (RT)
Expand
View Pricing
01-4870
Ammonium oxalate monohydrate,
JIS special grade, ≥99.5%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More