Ammonium dichromate
Synonym(s):
Ammonium bichromate, Ammonium dichromate
Linear Formula:
(NH4)2Cr2O7
CAS Number:
Molecular Weight:
252.06
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing