Skip to Content
MilliporeSigma
Acetic acid, glacial EMPROVE® EXPERT, Ph. Eur., JP, USP

Acetic acid (glacial)

Acetic acid, glacial EMPROVE® EXPERT, Ph. Eur., JP, USP
Synonym(s):
Acetic acid, Glacial acetic acid
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Technique