Skip to Content
MilliporeSigma
7-Hydroxycoumarin-3-carboxylic acid suitable for fluorescence, ≥98.0% (capillary electrophoresis)

7-Hydroxycoumarin-3-carboxylic acid

7-Hydroxycoumarin-3-carboxylic acid suitable for fluorescence, ≥98.0% (capillary electrophoresis)
Synonym(s):
Umbelliferone-3-carboxylic acid
Empirical Formula (Hill Notation):
C10H6O5
CAS Number:
Molecular Weight:
206.15

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
pK Value
Purity
Quality Segment
Showing 1-1 of 1