
4-tert-Butylphenol

Synonym(s):
4-tert-Butylphenol
Linear Formula:
(CH3)3CC6H4OH
CAS Number:
Molecular Weight:
150.22
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing