
4,4′-Methylenebis(2,6-di-tert-butylphenol)

Synonym(s):
4,4′-Methylenebis(2,6-di-tert-butylphenol)
Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing