
2-Undecanone

Synonym(s):
Methyl nonyl ketone, 2-Undecanone
Linear Formula:
CH3(CH2)8COCH3
CAS Number:
Molecular Weight:
170.29
Products
Shipping
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing