Skip to Content
MilliporeSigma
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine ≥95.5% (GC), ≥98% (TLC)

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine ≥95.5% (GC), ≥98% (TLC)
Synonym(s):
(7R,17Z)-4-Hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-3,5,8-trioxa-4-phosphahexacos-17-en-1-aminium 4-oxide, inner salt, 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 2-oleoyl-1-palmitoyl, L-β-Oleoyl-γ-palmitoyl-α-lecithin, PC(16:0/18:1(9Z)), PC(16:0/18:1), PC(16:0/18:1w9), POPC
Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Functional Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing