Skip to Content
Merck
2-Dimethylaminoethanol SAJ first grade, ≥99.0%

2-Dimethylaminoethanol

2-Dimethylaminoethanol SAJ first grade, ≥99.0%
Synonym(s):
N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA, 2-(Dimethylamino)-ethanol
Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6