
2-Chlorobenzoyl chloride

Synonym(s):
2-Chlorobenzoyl chloride
Linear Formula:
2-(Cl)C6H4COCl
CAS Number:
Molecular Weight:
175.01
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing