Key Documents
S916838
2,2-DICHLORO-2',3',5',6'-TETRAFLUOROACETANILIDE
AldrichCPR
Sign Into View Organizational & Contract Pricing
Select a Size
50MG
$30.70
$30.70
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
Select a Size
Change View
50MG
$30.70
About This Item
Linear Formula:
C8H3Cl2F4NO
CAS Number:
Molecular Weight:
276.019
MDL number:
$30.70
Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.