Key Documents
668680
Sigma-Aldrich
7-Methylindole-2-carboxylic acid
95%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
95%
mp
169-175 °C
SMILES string
Cc1cccc2cc([nH]c12)C(O)=O
InChI
1S/C10H9NO2/c1-6-3-2-4-7-5-8(10(12)13)11-9(6)7/h2-5,11H,1H3,(H,12,13)
InChI key
OLNYNTKNRVFVBI-UHFFFAOYSA-N
Application
  • Reactant for preparation of N-terminal anthranoyl-phenylalanine derivatives as CCK1 receptor antagonists
  • Reactant for preparation of factor Xa inhibitors based on a 2-carboxyindole scaffold
  • Reactant for preparation of potent and selective non-imidazole human histamine H4 receptor antagonists
pictograms
Exclamation mark
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)