Key Documents
459259
Sigma-Aldrich
(1R,2R)-(−)-trans-1,2-Cyclopentanediol
98%
Sign Into View Organizational & Contract Pricing
About This Item
Linear Formula:
C5H8(OH)2
CAS Number:
Molecular Weight:
102.13
Beilstein/REAXYS Number:
1901116
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
assay
98%
optical activity
[α]20/D −21°, c = 1 in chloroform
optical purity
ee: 97% (GLC)
mp
47-51 °C (lit.)
SMILES string
O[C@@H]1CCC[C@H]1O
Looking for similar products? Visit Product Comparison Guide
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Choose from one of the most recent versions:
Certificates of Analysis (COA)