Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000727

Sigma-Aldrich

Chromium(III) sulfate basic

puriss.

Synonym(s):

Chromosal B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr4(SO4)5(OH)2
CAS Number:
Molecular Weight:
722.31
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

puriss.

SMILES string

[Cr+3].[Cr+3].[Cr+3].O[Cr+]O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/4Cr.5H2O4S.2H2O/c;;;;5*1-5(2,3)4;;/h;;;;5*(H2,1,2,3,4);2*1H2/q4*+3;;;;;;;/p-12

InChI key

CHFNDYXSNLPPCO-UHFFFAOYSA-B

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service