Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000564

Sigma-Aldrich

Tin(IV) chloride pentahydrate

98.0%

Synonym(s):

Tin tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl4 · 5H2O
CAS Number:
Molecular Weight:
350.60
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98.0%

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Sn](Cl)(Cl)Cl

InChI

1S/4ClH.5H2O.Sn/h4*1H;5*1H2;/q;;;;;;;;;+4/p-4

InChI key

KHMOASUYFVRATF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monaam Ben Ali et al.
Journal of colloid and interface science, 457, 360-369 (2015-07-22)
Zinc stannate (Zn2SnO4 or ZTO) nanoparticles were synthesized via hydrothermal method using NaOH as a mineralizer. X-ray diffraction (XRD), transmission electron microscopy (TEM) and high-resolution transmission electron microscopy (HRTEM) of the synthesized ZTO nanoparticles revealed the formation of highly pure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service