Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS1044

Norflurazon

analytical standard

Synonym(s):

4-Chloro-5-(methylamino)-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9ClF3N3O
CAS Number:
Molecular Weight:
303.67
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1044

application(s)

agriculture
environmental

SMILES string

CNC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F

InChI

1S/C12H9ClF3N3O/c1-17-9-6-18-19(11(20)10(9)13)8-4-2-3-7(5-8)12(14,15)16/h2-6,17H,1H3

InChI key

NVGOPFQZYCNLDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geetanjali Saini et al.
The Plant journal : for cell and molecular biology, 65(5), 690-702 (2011-01-07)
Various mutant screens have been undertaken to identify constituents involved in the transmission of signals from the plastid to the nucleus. Many of these screens have been performed using carotenoid-deficient plants grown in the presence of norflurazon (NF), an inhibitor
Maneesha R Aluru et al.
Plant physiology, 150(2), 904-923 (2009-04-24)
Arabidopsis (Arabidopsis thaliana) immutans (im) has green and white sectoring due to the action of a nuclear recessive gene, IMMUTANS. The green sectors contain normal-appearing chloroplasts, whereas the white sectors contain abnormal chloroplasts that lack colored carotenoids due to a
Holger Nestler et al.
Aquatic toxicology (Amsterdam, Netherlands), 110-111, 214-224 (2012-02-24)
The release of herbicides into the aquatic environment raises concerns about potential detrimental effects on ecologically important non-target species, such as unicellular algae, necessitating ecotoxicological risk assessment. Algal toxicity tests based on growth, a commonly assessed endpoint, are integrative, and
Ke Sun et al.
Environmental pollution (Barking, Essex : 1987), 163, 167-173 (2012-02-14)
Sorption of two herbicides, fluridone (FLUN) and norflurazon (NORO), by two types of biochars, whole sediment, and various soil/sediment organic matter (OM) fractions including nonhydrolyzable carbon (NHC), black carbon (BC) and humic acid (HA) was examined. The single-point organic carbon
Ssu-Wei Hsu et al.
Plant & cell physiology, 51(7), 1197-1209 (2010-05-22)
Here, we report unique desiccation-associated ABA signaling transduction through which the Rop (Rho GTPase of plants) gene is regulated during the stage of pollen maturation. A gene encoding Rho GTPase was identified in lily (Lilium longiflorum Thunb.) pollen. Phylogenetic tree

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service