Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11409

Carbosulfan

analytical standard

Synonym(s):

2,3-Dihydro-2,2-dimethylbenzofuran-7-yl (dibutylaminothio)methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32N2O3S
CAS Number:
Molecular Weight:
380.54
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-1063

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCCCN(CCCC)SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C20H32N2O3S/c1-6-8-13-22(14-9-7-2)26-21(5)19(23)24-17-12-10-11-16-15-20(3,4)25-18(16)17/h10-12H,6-9,13-15H2,1-5H3

InChI key

JLQUFIHWVLZVTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Umetsu et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 15(1), 1-23 (1980-01-01)
The cleavage of the N-S bond in 2,3-dihydro-2,2-dimethyl-7-benzofuranyl (di-n-butylaminosulfenyl) (methyl)carbamate was examined in different buffer solutions (hydrolysis), in buffer solution containing sulfhydryl reagents (thiolysis) and on thin-layer chromatographic plates. In buffer solution and on thin-layer plates, N-S bond cleavage readily
Shuwen Wu et al.
Insect biochemistry and molecular biology, 65, 75-82 (2015-09-13)
Control of Chinese Apolygus lucorum relies heavily on organophosphate insecticides. Here we describe resistance to the organophosphate chlorpyrifos in an A. lucorum strain, BZ-R, which was developed from a field-collected strain (BZ) by selection with chlorpyrifos in the laboratory. BZ-R showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service