Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48612

Supelco

trans-1,2-Dichloroethene solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl2
CAS Number:
Molecular Weight:
96.94
EC Number:
UNSPSC Code:
23151816

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C2H2Cl2/c3-1-2-4/h1-2H/b2-1+

InChI key

KFUSEUYYWQURPO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Andrew Ramsburg et al.
Environmental science & technology, 44(23), 9105-9111 (2010-11-09)
Abiotic and biotic reductive dechlorination with chlorinated ethene dense non-aqueous-phase liquid (DNAPL) source zones can lead to significant fluxes of complete and incomplete transformation products. Accurate assessment of in situ rates of transformation and the potential for product sequestration requires
Darlene D Wagner et al.
BMC genomics, 13, 200-200 (2012-05-24)
Geobacter lovleyi is a unique member of the Geobacteraceae because strains of this species share the ability to couple tetrachloroethene (PCE) reductive dechlorination to cis-1,2-dichloroethene (cis-DCE) with energy conservation and growth (i.e., organohalide respiration). Strain SZ also reduces U(VI) to
Timothy E Mattes et al.
FEMS microbiology reviews, 34(4), 445-475 (2010-02-12)
Extensive use and inadequate disposal of chloroethenes have led to prevalent groundwater contamination worldwide. The occurrence of the lesser chlorinated ethenes [i.e. vinyl chloride (VC) and cis-1,2-dichloroethene (cDCE)] in groundwater is primarily a consequence of incomplete anaerobic reductive dechlorination of
He-Ping Zhao et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(9), 1796-1803 (2011-10-25)
Degradation of the lower chlorinated ethenes is crucial to the application of natural attenuation or in situ bioremediation on chlorinated ethene contaminated sites. Recently, within mixtures of several chloroethenes as they can occur in contaminated groundwater inhibiting effects on aerobic
He-Ping Zhao et al.
Water research, 44(7), 2276-2282 (2010-01-19)
The presence of other chloroethenes influences aerobic metabolic biodegradation of cis-1,2-dichloroethene (cDCE). A new metabolically cDCE degrading enrichment culture was identified as also being capable of degrading vinyl chloride (VC), but not 1,1-dichloroethene (1,1DCE), trans-1,2-dichloroethene (tDCE), trichloroethene (TCE), or tetrachloroethene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service