Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442503

Supelco

Benzyl butyl phthalate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[CH3(CH2)3O2C]C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
312.36
Beilstein/REAXYS Number:
2062204
EC Number:
MDL number:
UNSPSC Code:
41115711
PubChem Substance ID:

grade

analytical standard

vapor density

10.8 (vs air)

vapor pressure

0.16 mmHg ( 150 °C)

CofA

current certificate can be downloaded

autoignition temp.

450 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.54 (lit.)

density

1.1 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

Storage temp.

2-30°C

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCc2ccccc2

InChI

1S/C19H20O4/c1-2-3-13-22-18(20)16-11-7-8-12-17(16)19(21)23-14-15-9-5-4-6-10-15/h4-12H,2-3,13-14H2,1H3

Inchi Key

IRIAEXORFWYRCZ-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

General description

This product has been discontinued. Product 36927-250mg may be an acceptable replacement.

Pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113.0 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stir bar sorptive extraction and trace analysis of selected endocrine disruptors in water, biosolids and sludge samples by thermal desorption with gas chromatography-mass spectrometry.
Tan BLL, et al.
Water Research, 42(1-2), 404-412 (2008)
Rapid analysis of six phthalate esters in wine by ultrasound-vortex-assisted dispersive liquid-liquid micro-extraction coupled with gas chromatography-flame ionization detector or gas chromatography-ion trap mass spectrometry.
Cinelli G, et al.
Analytica Chimica Acta, 769(1-2), 72-78 (2013)
Gas chromatography-mass spectrometry determination of the migration of phthalate plasticisers from polyvinyl chloride toys and childcare articles.
Earls AO.
Journal of Chromatography A, 983(1-2), 237-246 (2003)
Determination of phthalates and adipate in bottled water by headspace solid-phase microextraction and gas chromatography/mass spectrometry.
Cao XL.
Journal of Chromatography A, 1178(1-2), 231-238 (2008)
Susan Sumner et al.
Journal of applied toxicology : JAT, 29(8), 703-714 (2009-09-05)
This study was conducted to evaluate the use of metabolomics for improving our ability to draw correlations between early life exposures and reproductive and/or developmental outcomes. Pregnant CD rats were exposed by gavage daily during gestation to vehicle or to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service