Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442378

Supelco

3,5-Dichlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein/REAXYS Number:
1906458
EC Number:
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

233 °C (lit.)

mp

65-68 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

Oc1cc(Cl)cc(Cl)c1

InChI

1S/C6H4Cl2O/c7-4-1-5(8)3-6(9)2-4/h1-3,9H

InChI key

VPOMSPZBQMDLTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Product 442378 is being discontinued for sale. Product 31595-250MG may be an acceptable replacement.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phenols and chlorophenols as trimethylsilyl derivatives using gas chromatography-mass spectrometry.
Kovacs A, et al.
Journal of Chromatography A, 1194(1), 139-142 (2008)
Evaluation of a dedicated gas chromatography-mass spectrometry method for the analysis of phenols in water.
Vermeulen A, et al.
Journal of Chromatography A, 1071(1-2), 41-46 (2005)
Development and optimization of a method for the analysis of phenols and chlorophenols from aqueous samples by gas chromatography-mass spectrometry, after solid-phase extraction and trimethylsilylation.
Kovacs A, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 99(1), 125-131 (2011)
Determination of chlorophenols by solid-phase microextraction and liquid chromatography with electrochemical detection.
Sarrion NM, et al.
Journal of Chromatography A, 947(2), 155-165 (2002)
Determination of chlorophenols in water samples using simultaneous dispersive liquid--liquid microextraction and derivatization followed by gas chromatography-electron-capture detection.
Fattahi N, et al.
Journal of Chromatography A, 1157(1-2), 23-29 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service