25879-U
Nitrogen (99.99%)
cylinder of 14 L, analytical standard
About This Item
Recommended Products
grade
analytical standard
form
gas
packaging
cylinder of 14 L
manufacturer/tradename
Scott Gas T102-14
availability
not available in EU, Canada
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
neat
Looking for similar products? Visit Product Comparison Guide
General description
Contents: 14 liters
Pressure: 240 psig
Outlet fitting: CGA-160-1/8 inch, NPT F
Weight: 1.5 lb.
Dimensions: 3 x 11 inches
D.O.T. specifications: 4B240
Application
Features and Benefits
- Nitrogen (99.99%) pure gas is suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
- A certificate of analysis is included.
- Packaged in lightweight, non-refillable, recyclable containers.
- Portable, convenient, and easy to use.
- Manufactured gravimetrically for maximum accuracy.
- NIST traceable.
regulator
signalword
Warning
hcodes
pcodes
Hazard Classifications
Press. Gas Compr. Gas
Storage Class
2A - Gases
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service