Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20639-U

Supelco

Oxisorb® Oxygen Scrubber

1/4 in. fitting

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115717

description

Kit

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

This trap will reduce oxygen and moisture to less than 1 ppm when the incoming level of oxygen is below 15 ppm and the incoming level of moisture is below 10 ppm. Use alternative traps when the levels in the incoming gas exceed these values. The kit includes a cartridge, and fittings for installation. Replacing the cartridge is simple, and can be performed quickly.

Dimensions:
  • Cartridge and Fittings: 91/4 in. (23.5 cm) long x 2 in. (51 mm) diameter
  • Cartridge Only: 47/8 in. (12.4 cm) long x 11/8 in. (29 mm) diameter

Application

N2, CO2 and CO2-N2 gas mixture may be passed through Oxisorb® Oxygen Scrubber before being used in an experimental study, done to monitor the kinetics of sulphate and acetate uptake, by the sulphate-reducing bacteria Desulfobacter postgatei.[1]

Legal Information

Oxisorb is a registered trademark of Messer Group GmbH

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

K Ingvorsen et al.
Applied and environmental microbiology, 47(2), 403-408 (1984-02-01)
The kinetics of sulfate and acetate uptake was studied in the sulfate-reducing bacterium Desulfobacter postgatei (DSM 2034). Kinetic parameters (K(m) and V(max)) were estimated from substrate consumption curves by resting cell suspensions with [S]sulfate and [C]acetate. Both sulfate and acetate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service