Key Documents
Z719927
MagaPhase® 96 Well Plate (Side-Pull) Magnetic Separator
About This Item
UNSPSC Code:
41100000
General description
MagaPhase® Magnetic Separation accessories consist of powerful magnetic devices designed to keep magnetizable beads in place while buffer is exchanged, or unwanted material is removed. These durable devices are engineered to accommodate industry standard microtiter plates and microfuge tubes for a wide range of magnetic separation applications including nucleic acid isolation, purification of biomolecules, as well as cell separation and immunoprecipitation using a pin magnet for a unique side-pull configuration.
Legal Information
MagaPhase is a registered trademark of Cortex Biochem, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)