Key Documents
Z622761
Ohaus® Analytical Plus balance
Ohaus® AP250D
About This Item
Pricing and availability is not currently available.
AC/DC input
220 V
feature
CE compliant
CSA approved
manufacturer/tradename
Ohaus® AP250D
availability
not available in USA and Canada
weighing capacity measuring range
250 g, resolution: 0.1 mg
platform diam.
90 mm
General description
Model AP250D offers delta ranging with 0.01mg resolution up to 52 g, and 0.1mg up to the full scales of 210g
The Analytical Plus series weigh in 8 different units, count pieces and have built-in GLP features.
  • RS232 interface
  • Lock switch
  • Internal calibration weight
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)