Z375071
Vertical electrophoresis unit, adjustable height, dual gel
gel W × H 16.5 cm × 14.5-28 cm, CE-approved
All Photos(1)
About This Item
Recommended Products
grade
electrophoresis grade
gel W × H
16.5 cm × 14.5-28 cm
Features and Benefits
- Made from high quality acrylic with platinum electrodes
- Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
- Safety covers with attached safety-tip power leads
- Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
- Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
- Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
- Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and four gel-mounting spring clamps.
CE-APPROVED ONLY WHERE INDICATED
Slab and tube gel accessories are not included; order from the accessories list
Choose from one of the most recent versions:
Certificates of Analysis (COA)