Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5451

Sigma-Aldrich

Tissue plasminogen activator human

pkg of 10 μg, ≥400,000 IU/mg (activity expressed per mg of tPA)

Synonym(s):

tPA

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in CHO cells

form

lyophilized powder

specific activity

≥400,000 IU/mg (activity expressed per mg of tPA)

mol wt

~70 kDa

packaging

pkg of 10 μg

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... PLAT(5327)

Looking for similar products? Visit Product Comparison Guide

Application

Tissue plasminogen activator has been used in a study to assess treatment risks of intracranial hemorrhage among patients with acute ischemic stroke. [1] Tissue plasminogen activator has also been used in a study to investigate the successful treatment of early thrombosis of HeartWare left ventricular assist devices with intraventricular thrombolytics. [2]

Physical form

Supplied as lyophilized powder containing 10 μg tPA lyophilized from a 100 μg/ml solution containing 1 M NH4HCO4 with 1% bovine serum albumin.
Tissue Plasminogen Activator (tPA) is a protease of the S1 family (trypsin family) and is found in a wide variety of mammalian tissues, especially endothelial cells. tPA is secreted as a single chain precursor, which is cleaved to a two-chain form by plasmin. Supplied in the single chain form with a molecular weight of ~70 kDa.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdallah Kamouh et al.
The Annals of thoracic surgery, 94(1), 281-283 (2012-06-28)
In the last few years, left ventricular assist devices (LVADs) have moved to the forefront in the management of patients with advanced heart failure. Pumps have gradually become smaller and more efficient and have clearly demonstrated survival benefits and improvement
Risks of intracranial hemorrhage among patients with acute ischemic stroke receiving warfarin and treated with intravenous tissue plasminogen activator
Xian, Y., et al.
Journal of the American Medical Association, 307, 2600-2608 (2012)
Dan Huang et al.
Frontiers of medicine, 11(3), 410-422 (2017-07-09)
Aberrant expression of annexin A2-S100A10 heterotetramer (AIIt) associated with PML/RARα fusion protein causes lethal hyperfibrinolysis in acute promyelocytic leukemia (APL), but the mechanism is unclear. To facilitate the investigation of regulatory association between ANXA2 and promyelocytic leukemia/retinoic acid receptor a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service