Skip to Content
Merck
All Photos(1)

Key Documents

T3132

Sigma-Aldrich

Thymolphthalein monophosphate magnesium salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H29O7PMg
CAS Number:
Molecular Weight:
532.80 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103

Pricing and availability is not currently available.

assay

≥97% (HPLC)

suitability

suitable for substrate for alkaline phosphatase

storage temp.

−20°C

SMILES string

[Mg++].CC(C)c1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(OP([O-])([O-])=O)cc4C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service