Skip to Content
MilliporeSigma
All Photos(1)

Documents

T3021

Sigma-Aldrich

Tinidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

SMILES string

CCS(=O)(=O)CCn1c(C)ncc1N(=O)=O

General description

Chemical structure: imidazole

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Federico et al.
Gastroenterology, 143(1), 55-61 (2012-04-10)
Helicobacter pylori have become resistant to antimicrobial agents, reducing eradication rates. A 10-day sequential regimen that contains levofloxacin was efficient, safe, and cost saving in eradicating H pylori infection in an area with high prevalence of clarithromycin resistance. We performed
Elizabeth A Bosserman et al.
Sexually transmitted diseases, 38(10), 983-987 (2011-09-22)
Antimicrobial resistance is one of the causes of treatment failure in women after standard nitroimidazole therapy for Trichomonas vaginalis infections. The Centers for Disease Control and Prevention provides drug susceptibility testing and guidance for treatment failures but the efficacy of
Hyuk Soon Choi et al.
World journal of gastroenterology, 18(19), 2377-2382 (2012-06-02)
To compare the effectiveness of sequential therapy for Helicobacter pylori (H. pylori) infection with that of triple therapy of varying durations. The 460 patients enrolled in this study had H. pylori-associated gastritis or a gastric or duodenal ulcer. After screening
C E Nord et al.
Infection, 11(1), 54-60 (1983-01-01)
Tinidazole is a 5-nitroimidazole with selective activity against anaerobic bacteria and protozoa. It is bactericidal at low concentrations and its spectrum covers most anaerobic bacteria and some capnophilic microorganisms. Anaerobic bacteria known to be resistant to tinidazole include anaerobic streptococci
Arlene C Seña et al.
The Journal of infectious diseases, 206(3), 357-365 (2012-05-23)
Chlamydia trachomatis (CT), Mycoplasma genitalium (MG), and Trichomonas vaginalis (TV) are sexually transmitted infections (STIs) associated with nongonococcal urethritis (NGU). We assessed their predictors and persistence after treatment. We analyzed data from an NGU treatment trial among symptomatic heterosexual men

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service