Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5319

Sigma-Aldrich

Sodium Stibogluconate

≥98% (titration), solid

Synonym(s):

2,4:2′,4′-O-(Oxy-distibylidyne)bis[D-gluconic acid] Sb,Sb-dioxide trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17O17Sb2 ·3Na
CAS Number:
Molecular Weight:
745.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (titration)

form

solid

storage condition

desiccated
protect from light
under inert gas

color

pale yellow

solubility

H2O: 1 mg/mL at ~75 °C

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H][C@@]1(O[SbH]([O-])(O[C@H]([C@H]1O)C([O-])=O)O[Sb]2(=O)O[C@H]([C@@H](O)[C@]([H])(O2)[C@H](O)CO)C([O-])=O)[C@H](O)CO

Biochem/physiol Actions

Irreversibly inhibits protein tyrosine phosphatase (SHP-1) activity (99% inhibition at 11 μM).
Irreversibly inhibits protein tyrosine phosphatase (SHP-1) activity (99% inhibition at 11μM). Inhibits SHP-2 and PTP1B at 10-folds higher concentration. Induces general tyrosine phosphorylation. Augments IL-3-induced proliferation in Baf3 cells (Emax=55 μM).

Packaging

Package under nitrogen, protected from light with a desiccant.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service