S4627
Stearyl stearate
~99%
Synonym(s):
Stearic acid stearyl ester
About This Item
Recommended Products
biological source
plant (vegetable)
assay
~99%
form
powder
functional group
ester
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC
InChI
1S/C36H72O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-35H2,1-2H3
InChI key
NKBWPOSQERPBFI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service