Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N8289

Sigma-Aldrich

NBI-31772 hydrate

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11NO7 · xH2O
CAS Number:
Molecular Weight:
341.27 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

storage condition

protect from light

storage temp.

−20°C

SMILES string

O.OC(=O)c1cc2cc(O)c(O)cc2c(n1)C(=O)c3ccc(O)c(O)c3

InChI

1S/C17H11NO7.H2O/c19-11-2-1-7(4-12(11)20)16(23)15-9-6-14(22)13(21)5-8(9)3-10(18-15)17(24)25;/h1-6,19-22H,(H,24,25);1H2

InChI key

QHAUFGIHMBUQKN-UHFFFAOYSA-N

Biochem/physiol Actions

NBI-31772 is an insulin-like growth factor-1 binding proteins (IGFBPs) inhibitor; IGF-1 potentiator.
NBI-31772 is an insulin-like growth factor-1 binding proteins (IGFBPs) inhibitor; IGF-1 potentiator. NBI-31772 is an air/DMSO DOPA oxidation product, which inhibits IGF binding to IFGFB′s. NBI-31772 displaces IGF-1 from IGF1:IGFBP complex. Thus, it increases circulating level of IGF-1. NBI-31772 does not bind to IGFR.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X J Liu et al.
The Journal of biological chemistry, 276(35), 32419-32422 (2001-07-11)
Insulin-like growth factor-I (IGF-I) has both metabolic and mitogenic activities mediated through interaction with the type 1 IGF receptor. The circulation of IGF-I in blood and interstitial fluid is not free but bound mostly to a family of six high
Jessica E Malberg et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 32(11), 2360-2368 (2007-03-08)
The present studies were conducted to determine if increasing central levels of the neurotrophic factor insulin-like growth factor-1 (IGF-I) either directly or indirectly produces anxiolytic and antidepressant-like effects in the mouse. Central levels of IGF-I can be increased directly, by
Jonathan D Schertzer et al.
The American journal of pathology, 171(4), 1180-1188 (2007-09-08)
Administration of recombinant human insulin-like growth factor-I (rhIGF-I) has beneficial effects in animal models of muscle injury and muscular dystrophy. However, the results of these studies may have been confounded by interactions of rhIGF-I with endogenous IGF-binding proteins (IGFBPs). To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service