Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N7769

Sigma-Aldrich

Nivalenol from Fusarium nivale

≥98%

Synonym(s):

3α,4β,7α,15-Tetrahydroxy-12,13-epoxytrichothec-9-en-8-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O7
CAS Number:
Molecular Weight:
312.32
MDL number:
UNSPSC Code:
12352200

assay

≥98%

storage temp.

2-8°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@H](O)[C@@H](O)[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Acute Tox. 2 Dermal

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deyu Huang et al.
Scientific reports, 7(1), 17079-17079 (2017-12-08)
Nivalenol (NIV), a type B trichothecenes commonly found in cereal crops, can cause growth impairment in animals. However, limited information about its mechanisms is available. Trichothecenes have been characterized as an inhibitor of protein synthesis and induce apoptosis in cells.
Jungkwan Lee et al.
Applied and environmental microbiology, 78(7), 2161-2167 (2012-01-31)
Fusarium graminearum (Gibberella zeae) is an important pathogen of wheat, maize, barley, and rice in South Korea, and harvested grain often is contaminated with trichothecenes such as deoxynivalenol and nivalenol. In this study, we examined 568 isolates of F. graminearum
Tadahiro Suzuki et al.
Journal of agricultural and food chemistry, 60(37), 9519-9527 (2012-08-18)
Type B trichothecenes, deoxynivalenol (DON) and nivalenol (NIV), are secondary metabolites of Fusarium species and are major pollutants in food and feed products. Recently, the production trend of their derivatives, 3-acetyldeoxynivalenol (3-AcDON), 15-acetyldeoxynivalenol (15-AcDON), and 4-acetylnivalenol (4-AcNIV or fusarenon-X), has
E D Van Asselt et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(10), 1556-1565 (2012-06-30)
Mycotoxins are secondary metabolites produced by fungi that can cause adverse health effects. Due to climate change, temperatures are expected to rise and changes in rainfall patterns are foreseen. These developments may increase fungal occurrence and mycotoxin concentrations in maize.
G Barros et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(2), 293-303 (2011-05-21)
Soybean (Glycine max L.), the main source of protein throughout the world, is used both as a food and a feedstuff. Currently, limited information about the occurrence of Fusarium species and mycotoxins in soybean grain and by-products is available. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service