Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK235

Sigma-Aldrich

Adenylate Kinase Activity Assay Kit (Colorimetric/Fluorometric)

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Adenylate Kinase Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

detection method

colorimetric
fluorometric

relevant disease(s)

hematological disorder; cardiovascular diseases; cancer

storage temp.

−20°C

General description

Adenylate Kinase (AK) (EC 2.7.4.3) is an abundant enzyme involved in energy metabolism and homeostasis of cellular adenine nucleotide ratios in different intracellular compartments. The enzyme is found in the nucleus, cytosol, or mitochondria (intermembrane space or matrix) of various kinds of tissues. Adenylate kinase acts on two molecules of ADP to generate ATP and AMP. Nine isoforms of adenylate kinase have been identified. Erythrocyte adenylate kinase deficiency is associated with hemolytic anemia. Adenylate kinase also plays an important role in post-ischemic recovery and in apoptosis.
About 30 – 40% of adenylate kinase activity takes place in mitochondria of heart muscles.[1] Mutation in adenylate kinase gene is associated with a severe combined immunodeficiency referred as reticular dysgenesis.[2]

Suitability

Suitable for the detection of Adenylate Kinase in cell/tissue homogenate, mitochondrial lysate, or purified protein.

Principle

This AK Activity Assay kit can kinetically measure Adenylate Kinase activity by detecting adenosine triphosphate (ATP) generated from adenosine diphosphate (ADP) as a substrate. ATP is detected via a multi-step reaction, resulting in the generation of an intermediate that reacts with the Adenylate Kinase Probe forming a colorimetric (570 nm)/fluorometric (ex = 535 nm/em = 587 nm) product.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Basis Of Congenital Neutropenia
Klein C.
The Hematology Journal : The Official Journal of the European Haematology Association / EHA (2009)
Adenylate kinase and AMP signaling networks: metabolic monitoring, signal communication and body energy sensing.
Dzeja P and Terzic A
International Journal of Molecular Sciences, 10(4) (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service