Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8253

Sigma-Aldrich

Maltotetraose

≥50% (HPLC)

Synonym(s):

Amylotetraose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
Beilstein/REAXYS Number:
78870
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

concentration

≥50% (HPLC)

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]4CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-5-9(29)10(30)15(35)22(40-5)44-19-7(3-27)42-24(17(37)12(19)32)45-20-8(4-28)41-23(16(36)13(20)33)43-18-6(2-26)39-21(38)14(34)11(18)31/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10+,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21+,22-,23-,24-/m1/s1

InChI key

LUEWUZLMQUOBSB-ZLBHSGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltotetraose is a maltooligosaccharide. [1]

Application

Maltotetraose producing amylases are highly desirable for application in bread making and other food industries. [1] It has been used in a study to characterize an exo-acting intracellular α-amylase from the hyperthermophilic bacterium Thermotoga neapolitana. [2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Kumar et al.
Bioresource technology, 116, 247-251 (2011-12-27)
Maltooligosaccharides especially maltotriose and maltotetraose producing amylases are highly desirable for application in bread making and other food industries. A maltotriose and maltotetraose producing amylase from moderately halophilic Marinobacter sp. EMB8 is described. Under optimized culture conditions, 48.0 IU/mL amylase
J Sauer et al.
Biochimica et biophysica acta, 1543(2), 275-293 (2001-01-11)
Glucoamylases are inverting exo-acting starch hydrolases releasing beta-glucose from the non-reducing ends of starch and related substrates. The majority of glucoamylases are multidomain enzymes consisting of a catalytic domain connected to a starch-binding domain by an O-glycosylated linker region. Three-dimensional
Carolyn Marion et al.
Infection and immunity, 79(10), 4193-4200 (2011-08-10)
Streptococcus pneumoniae is the leading cause of community-acquired pneumonia and results in over 1 million deaths each year worldwide. Asymptomatic colonization of the airway precedes disease, and acquisition of carbohydrates from the host environment is necessary for bacterial survival. We
Udo Schnupf et al.
Carbohydrate research, 344(3), 362-373 (2008-12-30)
DFT optimization studies of 90 syn alpha-maltotetraose (DP-4) amylose fragments have been carried out at the B3LYP/6-311++G** level of theory. The DP-4 fragments studied include V-helix, tightly bent conformations, a boat, and a (1)C(4) conformer. The standard hydroxymethyl rotamers (gg
Cécile Albenne et al.
FEBS letters, 527(1-3), 67-70 (2002-09-11)
Amylosucrase from Neisseria polysaccharea (AS) is a remarkable transglycosidase of family 13 of the glycoside hydrolases that catalyses the synthesis of an amylose-like polymer from sucrose and is always described as a sucrose-specific enzyme. Here, we demonstrate for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service