M7380
Myristyl acetate
liquid
Synonym(s):
Tetradecyl acetate
About This Item
form
liquid
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCOC(C)=O
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service