Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6629

Sigma-Aldrich

Ethyl heptadecanoate

Grade I, ≥99%

Synonym(s):

Heptadecanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
EC Number:
MDL number:
UNSPSC Code:
12352200

type

Grade I

assay

≥99%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service