Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G5260

Sigma-Aldrich

Dimethyl glutarate

≥98%

Synonym(s):

Glutaric acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCO(CH2)3COOCH3
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
1771444
EC Number:
MDL number:
UNSPSC Code:
12162002

assay

≥98%

refractive index

n20/D 1.424 (lit.)

bp

96-103 °C/15 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)CCCC(=O)OC

InChI

1S/C7H12O4/c1-10-6(8)4-3-5-7(9)11-2/h3-5H2,1-2H3

InChI key

XTDYIOOONNVFMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by D158801-ALDRICH | DBE-5 dibasic ester 99%

replaced by

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

228.2 °F - closed cup

flash_point_c

109.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service