Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G2750

Supelco

Gelsemine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O2
CAS Number:
Molecular Weight:
322.40
EC Number:
MDL number:
UNSPSC Code:
41116107

SMILES string

CN1C[C@]2(C=C)C3CC4OCC3C1[C@H]2[C@]45C(=O)Nc6ccccc56

InChI

1S/C20H22N2O2/c1-3-19-10-22(2)16-11-9-24-15(8-13(11)19)20(17(16)19)12-6-4-5-7-14(12)21-18(20)23/h3-7,11,13,15-17H,1,8-10H2,2H3,(H,21,23)/t11?,13?,15?,16?,17-,19-,20-/m0/s1

InChI key

NFYYATWFXNPTRM-ZIWPNRSCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

CNS stimulant from roots and rhizome of Gelsemium sempervirens

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service