Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F3764

Sigma-Aldrich

Furegrelate sodium salt

Synonym(s):

5-(3-Pyridinylmethyl)benzofurancarboxylic acid sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10NNaO3
CAS Number:
Molecular Weight:
275.23
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)c1cc2cc(Cc3cccnc3)ccc2o1

Gene Information

human ... TBXAS1(6916)

Biochem/physiol Actions

Thromboxane synthetase inhibitor.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - Solvent

flash_point_c

87 °C - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R R Gorman et al.
Prostaglandins, 26(2), 325-342 (1983-08-01)
This report outlines the activity of a new thromboxane synthase inhibitor sodium, 5-(3-pyridinylmethyl)-2-benzofurancarboxylate, (U-63557A). U-63557A is a potent inhibitor of the thromboxane synthase in human platelets in vitro, as well as in rhesus monkey platelets ex vivo. A single oral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service