Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

F0393

Sigma-Aldrich

2′-Fucosyl-D-lactose

from human milk, ≥98% (HPAE/PAD)

Synonym(s):

α-L-Fuc-(1→2)-β-D-Gal-(1→4)-D-Glc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O15
CAS Number:
Molecular Weight:
488.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

human milk

assay

≥98% (HPAE/PAD)

form

powder

solubility

H2O: 2 mg + 0.2 mL, clear, colorless

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C18H32O15/c1-4-7(21)9(23)13(27)17(29-4)33-15-10(24)8(22)5(2-19)31-18(15)32-14-6(3-20)30-16(28)12(26)11(14)25/h4-28H,2-3H2,1H3/t4-,5+,6+,7+,8-,9+,10-,11+,12+,13-,14+,15+,16?,17+,18-/m0/s1

InChI key

SNFSYLYCDAVZGP-ZTFPOLCTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Fucosyl-D-lactose is used as substrate to identify, differentiate and characterize fucosidases. It is also used in the development of biosensors to identify anti-infective oligosaccharides. 2′-Fucosyl-D-lactose is used to differentiate IL-8 receptor specificity and function. It may be used to study is effects on intestinal microbiota and as a reference compound in assays developed to detect and measure milk sugars.[1][2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myriam Coulet et al.
Regulatory toxicology and pharmacology : RTP, 68(1), 59-69 (2013-11-23)
In order to match the composition of human breast milk more closely, it is now possible to supplement commercial infant formula (IF) with synthesised oligosaccharides that are chemically identical to human milk oligosaccharides. The safety data generated on a new
C Campbell et al.
Cell, 35(1), 303-309 (1983-11-01)
Two rare and dominant mutants of Chinese hamster ovary (CHO) cells, LEC11 and LEC12, express the mouse embryonic antigen SSEA-1. Parental CHO cells and the revertants, LEC11.R9 and LEC12.R10, do not express this antigen as detected by a sensitive radioimmunoassay
M H Mansour et al.
Molecular and biochemical parasitology, 69(2), 173-184 (1995-02-01)
Two novel lectins that bind selectively to a schistosome-associated fucosyllactose-related determinant have been characterized and purified from the hemolymph of Biomphalaria alexandrina, the snail vector of Schistosoma mansoni. Both lectins were purified by affinity chromatography on a column of equimolar
K H Smalla et al.
Neuroreport, 9(5), 813-817 (1998-05-14)
Fucosylation of terminal galactose residues of brain glycoproteins in the alpha(1-2) position has been shown to be crucial for neuronal plasticity, including phenomena such as long-term potentiation and long-term memory formation. We raised antibodies against the plasticity-relevant fucalpha(1-2)gal epitope and
Jonathan A Lane et al.
Analytical biochemistry, 410(2), 200-205 (2010-11-30)
It is now well accepted that milk oligosaccharides can have a direct inhibitory effect on pathogenic microorganisms by interfering with their adhesion to human cells. Many free oligosaccharides from milk are considered to be soluble receptor analogs of epithelial cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service