Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

D115

Dextromethorphan hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C18H25NO · HBr
CAS Number:
Molecular Weight:
352.31
UNSPSC Code:
12352200
EC Number:
204-750-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

Br.COc1ccc2C[C@H]3C4CCCC[C@@]4(CCN3C)c2c1

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Nicholas Verne et al.
The journal of pain : official journal of the American Pain Society, 13(9), 901-909 (2012-09-11)
Irritable bowel syndrome (IBS) is a common gastrointestinal disorder in which the pathophysiological mechanisms of the pain and hypersensitivity are incompletely understood. IBS patients frequently complain of pain in body regions somatotopically distinct from the gut, suggesting involvement of central
Andreas Trojan et al.
Cancer chemotherapy and pharmacology, 71(2), 301-306 (2012-10-27)
Endoxifen serum concentrations seem to correlate with outcome in breast cancer (BC) patients. Concurrently, cytochrome P450 2D6 (CYP2D6) enzyme activity and dextromethorphan (DM) metabolism are deemed a surrogate marker for the formation of endoxifen. Here, we conducted a matched cohort
Shiou-Lan Chen et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 7(4), 1025-1033 (2012-09-20)
Recent studies show that proinflammatory cytokines might be related to the development of opioid dependence (physiological, psychological, or both). In a double-blind, randomly stratified clinical trial investigating whether add-on dextromethorphan (60-120 mg/day) attenuated inflammation and the combined use of opioids
Shinn-Jong Jiang et al.
Microcirculation (New York, N.Y. : 1994), 20(2), 190-201 (2012-11-13)
This study examines the effect of Dextromethorphan (d-3-methoxy-17-methylmorphinan; DXM), a commonly used cough-suppressing drug, on the expression of VCAM-1 and ICAM-1 in human umbilical vein endothelial cells (HUVECs) stimulated with lipopolysaccharide (LPS). The effect of DXM on expression of cell
F L Opdam et al.
Cancer chemotherapy and pharmacology, 71(3), 593-601 (2012-12-12)
Adjuvant therapy with tamoxifen significantly reduces breast cancer recurrence and mortality in estrogen receptor positive disease. CYP2D6 is the main enzyme involved in the activation of the prodrug tamoxifen into the anti-estrogen endoxifen. Endoxifen is thought to be a main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service