All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H14O8
CAS Number:
Molecular Weight:
346.29
EC Number:
UNSPSC Code:
12352200
Recommended Products
assay
90-95%
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Warning
Possibly carcinogenic.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service