Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7652

Sigma-Aldrich

D-Alanine benzyl ester p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H13NO2 · C7H8SO3
CAS Number:
Molecular Weight:
351.42
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.C[C@@H](N)C(=O)OCc2ccccc2

InChI

1S/C10H13NO2.C7H8O3S/c1-8(11)10(12)13-7-9-5-3-2-4-6-9;1-6-2-4-7(5-3-6)11(8,9)10/h2-6,8H,7,11H2,1H3;2-5H,1H3,(H,8,9,10)/t8-;/m1./s1

InChI key

NWOPHJSSBMABBD-DDWIOCJRSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service