Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96681

Sigma-Aldrich

Colupulone

mixture of homologues, ≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥85% (HPLC)

form

solid

storage temp.

−20°C

Biochem/physiol Actions

Colupulone is a bitter acid extracted from HOPS which activates transcription of xenobiotic metabolism via the human nuclear pregnane X receptor.
Mixture of predominantly colupulone and lupulone is an activator of the extrinsic apoptotic pathway via TRAIL DR4/DR5 death receptors and antimicrobial agent.

Analysis Note

predominantly colupulone and lupulone

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise G Teotico et al.
Molecular pharmacology, 74(6), 1512-1520 (2008-09-05)
Hops extracts are used to alleviate menopausal symptoms and as an alternative to hormone replacement therapy, but they can produce potentially harmful drug-drug interactions. The nuclear xenobiotic receptor pregnane X receptor (PXR) is promiscuously activated by a range of structurally
Steven A Kliewer et al.
Endocrine reviews, 23(5), 687-702 (2002-10-10)
The nuclear pregnane X receptor (PXR; NR1I2) is an important component of the body's adaptive defense mechanism against toxic substances including foreign chemicals (xenobiotics). PXR is activated by a large number of endogenous and exogenous chemicals including steroids, antibiotics, antimycotics
Steven A Kliewer et al.
Journal of lipid research, 43(3), 359-364 (2002-03-15)
The nuclear pregnane X receptor (PXR; NR1I2) is an integral component of the body's defense mechanism against chemical insult (chemoprotection). PXR is activated by a diverse array of lipophilic chemicals, including xenobiotics and endogenous substances, and regulates the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service