30927
2′-Deoxyadenosine 5′-triphosphate disodium salt
≥90% (HPLC)
Synonym(s):
dATP-Na2
About This Item
assay
≥90% (HPLC)
storage temp.
−20°C
SMILES string
[Na+].[Na+].Nc1ncnc2n(cnc12)C3C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3
InChI
1S/C10H16N5O12P3.2Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);;/q;2*+1/p-2/t5-,6+,7?;;/m0../s1
InChI key
JEKDCIBJADJZSK-LNUHRGGJSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service