Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25140

Sigma-Aldrich

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

purum, ≥97.0% (NMR)

Synonym(s):

N-Chloromethyl-N′-fluorotriethylenediammonium bis(tetrafluoroborate), F-TEDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14B2ClF9N2
CAS Number:
Molecular Weight:
354.26
Beilstein/REAXYS Number:
5368649
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

purum

assay

≥97.0% (NMR)

technique(s)

NMR: suitable (19F-, 1H-NMR)

mp

260 °C (lit.)

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2

InChI

1S/C7H14ClFN2.2BF4/c8-7-10-1-4-11(9,5-2-10)6-3-10;2*2-1(3,4)5/h1-7H2;;/q+2;2*-1

InChI key

TXRPHPUGYLSHCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for electrophilic fluorination [1][2][3]; Selective oxidizing agent[4]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.E. Banks et al.
Journal of the Chemical Society. Chemical Communications, 343-343 (1994)
D.P. Matthews et al.
Tetrahedron Letters, 34, 3057-3057 (1993)
L.G. Sankar
The Journal of Organic Chemistry, 58, 2791-2791 (1993)
R.E. Banks et al.
Synlett, 831-831 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service