Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17801

Sigma-Aldrich

Datiscetin

≥99.0% (HPLC)

Synonym(s):

2′,3,5,7-Tetrahydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
Beilstein/REAXYS Number:
39982
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0% (HPLC)

SMILES string

Oc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3ccccc3O

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aik Jiang Lau et al.
Pharmacological research, 100, 64-72 (2015-08-05)
Pregnane X receptor (PXR; NR1I2) is a member of the superfamily of nuclear receptors that regulates the expression of genes involved in various biological processes, including drug transport and biotransformation. In the present study, we investigated the effect of 3-hydroxyflavone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service